
Gonadorelin for system suitability

Synonym(s):
Gonadorelin
Empirical Formula (Hill Notation):
C55H75N17O13
Products
Formula Weight
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing