Formoterol fumarate for system suitability
Synonym(s):
Formoterol fumarate
Empirical Formula (Hill Notation):
C19H24N2O4 · 0.5C4H4O4
CAS Number:
Molecular Weight:
402.44
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing