
Fluram

Synonym(s):
Fluorescamine, 4-Phenylspiro-[furan-2(3H),1-phthalan]-3,3′-dione
Empirical Formula (Hill Notation):
C17H10O4
Products
Brand
Formula Weight
Melting Point (°C)
Product Line
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing