Endo-2-Naphthyl Kwon [2.2.1] Bicyclic Phosphine
Empirical Formula (Hill Notation):
C22H22NO2PS
Molecular Weight:
395.45
Products
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing