
Desloratadine for system suitability

Synonym(s):
Desloratadine, 8-Chloro-6,11-dihydro-11-(4-piperidinylidene)-5H- benzo[5,6]cyclohepta[1,2,b]pyridine, Clarinex®, NSC 675447
Empirical Formula (Hill Notation):
C19H19ClN2
CAS Number:
Molecular Weight:
310.82
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing