Skip to Content
TH
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
CdS/ZnS core-shell type quantum dots
Products
Product Category
quantum dots (8)
Brand
Sigma-Aldrich (8)
Physical Form
Functional Group
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
900308
CdS/ZnS core-shell type quantum dots,
lyophilized, fluorescence λ
em
425 nm, solid
Expand
View Pricing
900310
CdS/ZnS core-shell type quantum dots,
lyophilized, fluorescence λ
em
400 nm, solid
Expand
View Pricing
900304
CdS/ZnS core-shell type quantum dots,
amine functionalized, fluorescence λ
em
450 nm, 4 μM in PBS
Expand
View Pricing
900334
CdS/ZnS core-shell type quantum dots,
oleic acid functionalized, fluorescence λ
em
450 nm, 5 mg/mL in toluene
Expand
View Pricing
900332
CdS/ZnS core-shell type quantum dots,
lyophilized, fluorescence λ
em
450 nm, solid
Expand
View Pricing
900283
CdS/ZnS core-shell type quantum dots,
oleic acid functionalized, fluorescence λ
em
425 nm, solid
Expand
View Pricing
900301
CdS/ZnS core-shell type quantum dots,
PEG functionalized, fluorescence λ
em
450 nm, 4 μM in H
2
O
Expand
View Pricing
900294
CdS/ZnS core-shell type quantum dots,
carboxylic acid functionalized, fluorescence λ
em
450 nm, 1 mg/mL in H
2
O
Expand
View Pricing