Skip to Content
Merck
Bismuth nitrate oxide for spectrophotometric det., Bi, 71-74% Bi basis (KT)

Bismuth nitrate oxide

Bismuth nitrate oxide for spectrophotometric det., Bi, 71-74% Bi basis (KT)
Synonym(s):
Bismuth oxynitrate, Bismuth(III) nitrate basic, Bismuthyl nitrate
Linear Formula:
Bi(O)NO3
CAS Number:
Molecular Weight:
286.98
EC Number:

Products

Brand
Formula Weight
Physical Form
Purity
Application
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing