Skip to Content
Merck
Benzonase® Nuclease HC, Purity > 90%

Benzonase® Nuclease HC, Purity > 90%

Benzonase® Nuclease HC, Purity > 90%
Synonym(s):
Endonuclease from Serratia marcescens
CAS Number:

Products

Product Category
Brand
Biological Source
Physical Form
Recombinant Host
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing