
Benserazide for peak identification

Synonym(s):
Benserazide hydrochloride, DL-Serine 2-(2,3,4-trihydroxybenzyl)hydrazide hydrochloride
Empirical Formula (Hill Notation):
C10H15N3O5 · HCl
CAS Number:
Molecular Weight:
293.70
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing