Anti-ATM antibody,Mouse monoclonal
Synonym(s):
Anti-Ataxia-Telangiectasia Mutated, Monoclonal Anti-ATM antibody produced in mouse
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Recombinant Host
Antibody Form
Research Area
Conjugate
Quality Segment
Package Size
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing