![α-(4-Chlorophenyl)-α-[2-(dimethylamino)ethyl]-2-pyridineacetonitrile hydrochloride pharmaceutical impurity standard](/deepweb/assets/sigmaaldrich/product/structures/392/296/e82cdabb-0a1c-4298-b1e1-da9ab739168e/640/e82cdabb-0a1c-4298-b1e1-da9ab739168e.png)
α-(4-Chlorophenyl)-α-[2-(dimethylamino)ethyl]-2-pyridineacetonitrile hydrochloride
![α-(4-Chlorophenyl)-α-[2-(dimethylamino)ethyl]-2-pyridineacetonitrile hydrochloride pharmaceutical impurity standard](/deepweb/assets/sigmaaldrich/product/structures/392/296/e82cdabb-0a1c-4298-b1e1-da9ab739168e/640/e82cdabb-0a1c-4298-b1e1-da9ab739168e.png)
Synonym(s):
(2RS)-2-(4-Chlorophenyl)-4-dimethylamino-2-(2-pyridyl)butyronitril hydrochloride, Chlorpheniramine nitrile hydrochloride
Empirical Formula (Hill Notation):
C17H18ClN3 · HCl
Molecular Weight:
336.26
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing