
7-O-Methylwogonin

Synonym(s):
5-Hydroxy-7,8-dimethoxy-2-phenyl-4-chromenone, 5-Hydroxy-7,8-dimethoxy-2-phenyl-4H-1-benzopyran-4-one, 5-Hydroxy-7,8-dimethoxyflavone, 7,8-Dimethoxynorwogonin, Moslosooflavone
Empirical Formula (Hill Notation):
C17H14O5
CAS Number:
Molecular Weight:
298.29
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing