Skip to Content
Merck
7,16-Dibenzyl-1,4,10,13-tetraoxa-7,16-diazacyclooctadecane 97%

7,16-Dibenzyl-1,4,10,13-tetraoxa-7,16-diazacyclooctadecane

7,16-Dibenzyl-1,4,10,13-tetraoxa-7,16-diazacyclooctadecane 97%
Synonym(s):
1,10-Dibenzyl-1,10-diaza-18-crown-6, N,N′-Dibenzyl-4,13-diaza-18-crown-6
Empirical Formula (Hill Notation):
C26H38N2O4
CAS Number:
Molecular Weight:
442.59

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing