Skip to Content
Merck
CHAPS Vetec™, reagent grade, 96%

CHAPS

CHAPS Vetec™, reagent grade, 96%
Synonym(s):
CHAPS, 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate
Empirical Formula (Hill Notation):
C32H58N2O7S
CAS Number:
Molecular Weight:
614.88

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
Technique
Showing 1-5 of 5