Skip to Content
Merck
4-tert-Amylphenol 99%

4-tert-Amylphenol

4-tert-Amylphenol 99%
Linear Formula:
C2H5C(CH3)2C6H4OH
CAS Number:
Molecular Weight:
164.24
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing