
4-(Dimethylamino)pyridine solution

Synonym(s):
DMAP solution
Empirical Formula (Hill Notation):
C7H10N2
CAS Number:
Molecular Weight:
122.17
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing