Skip to Content
Merck
3-(Trifluoromethyl)pyrazole 99%

3-(Trifluoromethyl)pyrazole

3-(Trifluoromethyl)pyrazole 99%
Empirical Formula (Hill Notation):
C4H3F3N2
CAS Number:
Molecular Weight:
136.08

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing