Skip to Content
Merck
3-(Ethoxydimethylsilyl)propylamine 97%

3-(Ethoxydimethylsilyl)propylamine

3-(Ethoxydimethylsilyl)propylamine 97%
Linear Formula:
CH3CH2OSi(CH3)2(CH2)3NH2
CAS Number:
Molecular Weight:
161.32

Products

Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing