
3-(Dansylamino)phenylboronic acid

Synonym(s):
3-(5-Dimethylaminonaphthalene-1-sulfonylamino)benzeneboronic acid, 3-(Dansylamido)benzeneboronic acid
Empirical Formula (Hill Notation):
C18H19BN2O4S
Products
Brand
Formula Weight
Product Line
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing