3,4,4` Trichlorocarbanilide (Triclocarban)
Synonym(s):
3,4,4′-Trichlorocarbanilide, 1-(4-Chlorophenyl)-3-(3,4-dichlorophenyl)urea, TCC, Triclocarban
Linear Formula:
Cl2C6H3NHCONHC6H4Cl
CAS Number:
Molecular Weight:
315.58
Products
Brand
Formula Weight
Melting Point (°C)
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing