Skip to Content
Merck
Mitomycin C, Streptomyces caespitosus

Mitomycin C

Mitomycin C, Streptomyces caespitosus
Synonym(s):
Mitomycin C, Mitomycin C, Streptomyces caespitosus, Carrier-Free, Mitomycin C, Streptomyces caespitosus
Empirical Formula (Hill Notation):
C15H18N4O5
CAS Number:
Molecular Weight:
334.33

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Activity Spectrum
Quality Segment
Mode of Action