2-Butanone peroxide
Synonym(s):
Ethyl methyl ketone peroxide, Methyl ethyl ketone peroxide
Linear Formula:
C2H5C(OOH)(CH3)OOC(OOH)(CH3)C2H5
CAS Number:
Molecular Weight:
210.22
Products
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing