
1-(tert-Butyldimethylsilyl)imidazole

Synonym(s):
TBDMSIM
Empirical Formula (Hill Notation):
C9H18N2Si
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing