Skip to Content
Merck
(1R,2R)-trans-1,2-Cyclohexanediol 99%

(1R,2R)-trans-1,2-Cyclohexanediol

(1R,2R)-trans-1,2-Cyclohexanediol 99%
Synonym(s):
(1R)-trans-1,2-Cyclohexanediol
Linear Formula:
C6H10(OH)2
CAS Number:
Molecular Weight:
116.16

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing