![1,5-Isoquinolinediol ≥98% (HPLC), powder](/deepweb/assets/sigmaaldrich/product/structures/259/644/f08004f2-4490-400c-8aea-0684598c56f3/640/f08004f2-4490-400c-8aea-0684598c56f3.png)
1,5-Isoquinolinediol
![1,5-Isoquinolinediol ≥98% (HPLC), powder](/deepweb/assets/sigmaaldrich/product/structures/259/644/f08004f2-4490-400c-8aea-0684598c56f3/640/f08004f2-4490-400c-8aea-0684598c56f3.png)
Synonym(s):
1,5-Dihydroxyisoquinoline, 5-Hydroxy-1(2H)-isoquinolinone, DiQ
Empirical Formula (Hill Notation):
C9H7NO2
CAS Number:
Molecular Weight:
161.16
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing