15-Acetyldeoxynivalenol
Synonym(s):
15-O-Acetyl-4-deoxynivalenol from Fusarium graminearum, 15-Acetoxy-3α,7α-dihydroxy-12,13-epoxytrichothec-9-en-8-one
Empirical Formula (Hill Notation):
C17H22O7
CAS Number:
Molecular Weight:
338.35
Products
Product Category
Brand
Formula Weight
Format
Application
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing