
1,3,5-Cyclohexanetricarboxylic acid

Synonym(s):
cis,cis-1,3,5-Cyclohexanetricarboxylic acid
Linear Formula:
C6H9(CO2H)3
CAS Number:
Molecular Weight:
216.19
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Markush Class
Markush Group
Monomer Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing