Skip to Content
Merck
1,1-Dichloro-2,2-bis(4-chlorophenyl)ethene 99%

1,1-Dichloro-2,2-bis(4-chlorophenyl)ethene

1,1-Dichloro-2,2-bis(4-chlorophenyl)ethene 99%
Synonym(s):
4,4′-DDE
Linear Formula:
(ClC6H4)2C=CCl2
CAS Number:
Molecular Weight:
318.03
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing