Merck
  • Enhanced tumorigenesis and lymphatic metastasis of CD133+ hepatocarcinoma ascites syngeneic cell lines mediated by JNK signaling pathway in vitro and in vivo.
Enhanced tumorigenesis and lymphatic metastasis of CD133+ hepatocarcinoma ascites syngeneic cell lines mediated by JNK signaling pathway in vitro and in vivo.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2013-04-16)
Yanling Jin, Jun Mao, Huanxi Wang, Zhenhuan Hou, Wei Ma, Jun Zhang, Bo Wang, Yuhong Huang, Shizhu Zang, Jianwu Tang, Lianhong Li
ABSTRACT
Cancer stem cells (CSCs), stem-like cells, or tumor-initiating cells (TICs) may initiate tumorigenesis and metastasis, but neither the basic cell biology of CSCs nor the mechanisms of CSC-mediated tumor growth and lymphoid node metastasis are understood. Evidence suggests that CSC phenotype is maintained, at least in part, by altered JNK signaling. In this study, factors influencing the growth and metastatic potential of CSCs were examined by comparing CD133 surface antigen expression, proliferation, clonogenicity, invasive capacity, tumorigenicity, and expression of JNK-associated signaling molecules between the highly metastatic mouse hepatocarcinoma ascites syngeneic cell line Hca-F and the low metastasis potential line Hca-P. The Hca-F line exhibited higher clonogenic, proliferative, and invasive capacities than Hca-P cells, and a greater proportion of Hca-F cells were CD133 positive. In both cell lines, the CD133+ subpopulation showed significantly enhanced tumorigenicity and metastatic potential. An in vivo tumorigenicity assay in nude mice indicated that Hca-F cells possessed significantly higher tumorigenicity than Hca-P cells as indicated by larger tumors after inoculation. Expression levels of E-cadherin (CDH1), annexin VII, and JNK1 proteins were inversely correlated with CD133 expression in both Hca-F and Hca-P cells. These results demonstrate that CD133+ subpopulations of both Hca-F and Hca-P lines show CSC-like properties. However, Hca-F cells showed greater tumorigenicity and invasiveness, consistent with greater lymphatic metastasis capacity. We propose that tumorigenesis and lymphatic metastasis are regulated by JNK/P53/annexin VII and JNK/ATF-2/CDH1/annexin VII signal transduction pathways.