Merck
  • Transcriptome Profiling of the Dorsomedial Prefrontal Cortex in Suicide Victims.
Transcriptome Profiling of the Dorsomedial Prefrontal Cortex in Suicide Victims.
International journal of molecular sciences (2022-07-10)
Fanni Dóra, Éva Renner, Dávid Keller, Miklós Palkovits, Árpád Dobolyi
ABSTRACT
The default mode network (DMN) plays an outstanding role in psychiatric disorders. Still, gene expressional changes in its major component, the dorsomedial prefrontal cortex (DMPFC), have not been characterized. We used RNA sequencing in postmortem DMPFC samples to investigate suicide victims compared to control subjects. 1400 genes differed using log2FC > ±1 and adjusted p-value < 0.05 criteria between groups. Genes associated with depressive disorder, schizophrenia and impaired cognition were strongly overexpressed in top differentially expressed genes. Protein-protein interaction and co-expressional networks coupled with gene set enrichment analysis revealed that pathways related to cytokine receptor signaling were enriched in downregulated, while glutamatergic synaptic signaling upregulated genes in suicidal individuals. A validated differentially expressed gene, which is known to be associated with mGluR5, was the N-terminal EF-hand calcium-binding protein 2 (NECAB2). In situ hybridization histochemistry and immunohistochemistry proved that NECAB2 is expressed in two different types of inhibitory neurons located in layers II-IV and VI, respectively. Our results imply extensive gene expressional alterations in the DMPFC related to suicidal behavior. Some of these genes may contribute to the altered mental state and behavior of suicide victims.
Sigma-Aldrich
Anti-S-100 Protein Antibody, clone 15E2E2, clone 15E2E2, Chemicon®, from mouse