Merck
  • Deficits in pattern separation and dentate gyrus proliferation after rodent lateral fluid percussion injury.
Deficits in pattern separation and dentate gyrus proliferation after rodent lateral fluid percussion injury.
IBRO neuroscience reports (2021-04-17)
Erika A Correll, Benjamin J Ramser, Maxon V Knott, Robert E McCullumsmith, Jennifer L McGuire, Laura B Ngwenya
ABSTRACT
It has been demonstrated that adult born granule cells are generated after traumatic brain injury (TBI). There is evidence that these newly generated neurons are aberrant and are poised to contribute to poor cognitive function after TBI. Yet, there is also evidence that these newly generated neurons are important for cognitive recovery. Pattern separation is a cognitive task known to be dependent on the function of adult generated granule cells. Performance on this task and the relation to dentate gyrus dysfunction after TBI has not been previously studied. Here we subjected Sprague Dawley rats to lateral fluid percussion injury or sham and tested them on the dentate gyrus dependent task pattern separation. At 2 weeks after injury, we examined common markers of dentate gyrus function such as GSK3ß phosphorylation, Ki-67 immunohistochemistry, and generation of adult born granule cells. We found that injured animals have deficits in pattern separation. We additionally found a decrease in proliferative capacity at 2 weeks indicated by decreased phosphorylation of GSK3ß and Ki-67 immunopositivity as compared to sham animals. Lastly we found an increase in numbers of new neurons generated during the pattern separation task. These findings provide evidence that dentate gyrus dysfunction may be an important contributor to TBI pathology.
Product Description
Roche
Anti-Bromodeoxyuridine, from mouse IgG1 (clone: BMC9318)