Merck
  • Unexpected positive patch test reactions to sesquiterpene lactones in patients sensitized to the glucose sensor FreeStyle Libre.
Unexpected positive patch test reactions to sesquiterpene lactones in patients sensitized to the glucose sensor FreeStyle Libre.
Contact dermatitis (2019-06-06)
Anne Herman, Martin Mowitz, Olivier Aerts, Jeroen Pyl, Laurence de Montjoye, An Goossens, Magnus Bruze, Marie Baeck
ABSTRACT
Most diabetic patients sensitized to FreeStyle Libre react to isobornyl acrylate (IBOA), with a considerable number of them also showing unexpected positive patch test reactions to sesquiterpene lactone (SL) mix (SLM) tested in the baseline series. To compile patch test results of subjects affected, and provide potential explanations for this association. Fifty-three Freestyle Libre-allergic patients were patch tested with IBOA and/or SLM, and several were also patch tested with the components of SLM. Chromatographic analyses were performed on the glucose sensor, IBOA, and the components of SLM. Thirty-three patients reacted positively to the components of SLM, and 11 of 27 patients reacted positively to alantolactone, in particular. Gas chromatography-mass spectrometry (GC-MS) analyses did not detect these chemicals in the different parts of the glucose sensor, or in IBOA. Significant co-sensitizations between SLs on the one hand and the glucose sensor FreeStyle Libre and/or isobornyl acrylate on the other hand exist, without evidence of the presence of SLs via GC-MS analysis. Cross-reactions between them seem improbable. As a possible hypothesis, a common precursor for both, such as camphene, may exist.
Sigma-Aldrich
Costunolide, ≥97% (HPLC)