- Morphological profile determines the frequency of spontaneous calcium events in astrocytic processes.
Morphological profile determines the frequency of spontaneous calcium events in astrocytic processes.
Astrocytes express a complex repertoire of intracellular Ca2+ transients (events) that represent a major form of signaling within individual cells and in astrocytic syncytium. These events have different spatiotemporal profiles, which are modulated by neuronal activity. Spontaneous Ca2+ events appear more frequently in distal astrocytic processes and independently from each other. However, little is known about the mechanisms underlying such subcellular distribution of the Ca2+ events. Here, we identify the initiation points of the Ca2+ events within the territory of single astrocytes expressing genetically encoded Ca2+ indicator GCaMP2 in culture or in hippocampal slices. We found that most of the Ca2+ events start in an optimal range of thin distal processes. Our mathematical model demonstrated that a high surface-to-volume of the thin processes leads to increased amplitude of baseline Ca2+ fluctuations caused by a stochastic opening of Ca2+ channels in the plasma membrane. Suprathreshold fluctuations trigger Ca2+ -induced Ca2+ release from the Ca2+ stores by activating inositol 1,4,5-trisphosphate (IP3 ) receptors. In agreement with the model prediction, the spontaneous Ca2+ events frequency depended on the extracellular Ca2+ concentration. Astrocytic depolarization by high extracellular K+ increased the frequency of the Ca2+ events through activation of voltage-gated Ca2+ channels in cultured astrocytes. Our results suggest that the morphological profile of the astrocytic processes is responsible for tuning of the Ca2+ events frequency. Therefore, structural plasticity of astrocytic processes can be directly translated into changes in astrocytic Ca2+ signaling. This may be important for both physiological and pathological astrocyte remodeling.