Skip to Content
Merck
L-Glutamic acid NIST® RM 8574, USGS41

L-Glutamic acid

L-Glutamic acid NIST® RM 8574, USGS41
Synonym(s):
(S)-2-Aminopentanedioic acid, Glu, L-Glutamic acid
Linear Formula:
HO2CCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
147.13

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Agency
Format
Analyte Suitability
Quality Segment
Application
Optical Isomer
API Family
Reference Material Type
Special Grade
Technique