Starch from corn
Synonym(s):
Corn starch, Starch from maize, Maize MIR162, GMO Standard ERM-BF446
Linear Formula:
(C6H10O5)n
CAS Number:
Products
Product Category
Brand
Biological Source
Manufacturer
Agency
Format
Analyte Suitability
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing