Skip to Content
Merck
Vitamin E water-miscible 1 ml = 500 mg DL-a-tocopherol acetate for oral use, EMPROVE® ESSENTIAL, USP

Vitamin E

Vitamin E water-miscible 1 ml = 500 mg DL-a-tocopherol acetate for oral use, EMPROVE® ESSENTIAL, USP
Synonym(s):
(±)-α-Tocopherol, DL-all-rac-α-Tocopherol, Vitamin E
Empirical Formula (Hill Notation):
C29H50O2
CAS Number:
Molecular Weight:
430.71

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Application
Markush Class
Markush Group
Showing 1-1 of 1