DAPI ready made solution
Synonym(s):
4′,6-Diamidino-2-phenylindole dihydrochloride, 2-(4-Amidinophenyl)-6-indolecarbamidine dihydrochloride, DAPI dihydrochloride
Empirical Formula (Hill Notation):
C16H15N5 · 2HCl
CAS Number:
Molecular Weight:
350.25
Products
Product Category
Brand
Formula Weight
Physical Form
Detection Method
Quality Segment
Markush Class
Markush Group
Technique
Showing 1-2 of 2