Skip to Content
Merck
(R)-(+)-Glycidol 97%, optical purity ee: 98% (GLC)

(R)-(+)-Glycidol

(R)-(+)-Glycidol 97%, optical purity ee: 98% (GLC)
Synonym(s):
(R)-(+)-Oxirane-2-methanol
Empirical Formula (Hill Notation):
C3H6O2
CAS Number:
Molecular Weight:
74.08
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing