Skip to Content
Merck
Gly-Gly BioXtra, ≥99.0% (NT)

Gly-Gly

Gly-Gly BioXtra, ≥99.0% (NT)
Synonym(s):
Diglycine, Glycyl-glycine, Gly-Gly, Glycylglycine, Free Base
Linear Formula:
NH2CH2CONHCH2COOH
CAS Number:
Molecular Weight:
132.12
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Melting Point (°C)
pH Value
Physical Form
pK Value
Product Line
Purity
Quality Segment
Application
Special Grade
Technique
Showing 1-6 of 6