Skip to Content
Merck
EmbryoMax® Filtered Light Mineral Oil The EmbryoMax Filtered Light Mineral Oil is available in a 100 mL format and may be used for routine mouse embryonic stem cell culture applications.

Mineral oil

EmbryoMax® Filtered Light Mineral Oil The EmbryoMax Filtered Light Mineral Oil is available in a 100 mL format and may be used for routine mouse embryonic stem cell culture applications.
Synonym(s):
Standard oil, Mineral oil
CAS Number:

Products

Product Category
Brand
Color
Feature
Manufacturer
Physical Form
Product Line
Agency
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique