L-α-Phosphatidylcholine
Synonym(s):
1,2-Diacyl-sn-glycero-3-phosphocholine, 3-sn-Phosphatidylcholine, L-α-Lecithin, Azolectin, PC, L-α-Lecithin, Soybean, L-α-Lecithin, Egg Yolk, Highly Purified
CAS Number:
Products
Product Category
Brand
Biological Source
Color
Physical Form
Purity
Research Area
Tissue
Quality Segment
Functional Group
Application
Optical Isomer
Showing 1-13 of 13
Compare
Product Number
Description
Pricing