Merck
HomePrimary Cell Culture TechniquesADCC Assay Using BioIVT Cryopreserved NK Cells
ADCC Assay Using BioIVT Cryopreserved NK Cells
Antibody-dependent Cellular Cytotoxicity (ADCC)
Antibody-dependent cellular cytotoxicity (ADCC) is a cell-mediated immune assay commonly used for therapeutic drug discovery. The mechanism of ADCC can be divided into three steps (Figure 1):
  1. Specific antigens on the surface of the target cell are recognized and bound by antibody.
  2. The immune cell (effector cell) will recognize and bind to the antigen-bound antibody.
  3. The effector cell will trigger death of the target cell.
Assay principle for ADCC assay
Figure 1.Mechanism of ADCC
In general, natural killer (NK) cells, which can be purified from peripheral blood cells, are used as effector cells. Cryopreserved NK cells provide a more flexible, off-the-shelf solution for researchers performing ADCC assays.
In this study, we explored the use of cryopreserved NK cells in ADCC assays using luciferase-expressing A549 cells as target cells. A human anti-EGFR recombinant antibody was used to bind EGFR on the surface of the target cell and mediate cell death. Cell killing was determined by measuring the change in luciferase activity of target cells.
ADCC Protocol
A brief workflow for the ADCC assay using cryopreserved NK effector cells is show in Figure 2. For the complete step-by-step protocol, including tips and tricks for performing the assay with cryopreserved cells, materials, and assay optimizations, please refer to our ADCC protocol using cryopreserved NK effector cells.
ADCC protocol
Figure 2.ADCC Protocol
ADCC Assays Using Cryopreserved NK Effector Cells
Cryopreserved NK cells were incubated with luciferase-expressing A549 target cells at E:T ratios of 6:1 and 18:1 in the presence of increasing concentrations of anti-EGFR antibody over a 24-hour period. Experiments were performed in duplicate (Figure 3). The cryopreserved NK effector cells exhibited robust target cell killing in the ADCC assay when using a 24-hour timepoint (43.8% cell killing using an E:T ratio of 6:1 and 76.6% cell killing using an E:T ratio of 18:1, when live cell numbers are interpolated from a standard curve). Calculated EC50 for anti-EGFR antibody in the ADCC assay was 1.5 ng/mL at an E:T ratio of 6:1 and 2.5 ng/mL at an E:T ratio of 18:1.
ADCC assay using A549 target cells and cryopreserved NK effector cells
Figure 3.ADCC assay using cryopreserved NK cells as effector cells, A549 cells expressing a luciferase reporter gene as target cells, and increasing concentrations of anti-EGFR antibody.
E:T ratio
Calculated results
Target cell killing
6:1
43.8%
18:1
76.6%
EC50 (anti-EGFR antibody)
6:1
1.5 ng/mL
18:1
2.5 ng/mL
To determine reproducibility of the assay using cryopreserved NK effector cells, results were compared across three different experiments using NK cells from different vials, all provided from the same donor, for each experiment. Data indicated the assay was reproducible, with similar percentages of cell killing at each E:T ratio and similar EC50 values for anti-EGFR antibody across all ADCC assays (Figure 4).
ADCC assay reproducibility using A549 target cells and cryopreserved NK effector cells
Figure 4.Reproducibility of ADCC assay using cryopreserved NK effector cells. Similar target cell killing percentages and EC50 values for anti-EGFR antibody were observed at each E:T ratio.
E:T ratio
Experiment A
Experiment B
Experiment C
Target cell killing
6:1
43.8%
45.8%
50.6%
18:1
76.6%
61.7%
54.2%
EC50 (anti-EGFR antibody)
6:1
1.5 ng/mL
2.0 ng/mL
0.9 ng/mL
18:1
2.5 ng/mL
1.8 ng/mL
3.0 ng/mL
To determine donor variability, three lots of cryopreserved NK cells from three different donors were tested in the ADCC assay. The NK cells from all donors exhibited >50% cell killing using an E:T ratio of 18:1. Similar percentages of target cell killing were observed at each E:T ratio. Calculated EC50 values for anti-EGFR antibody were similar across all ADCC assays (Figure 5).
ADCC assay reproducibility using different donors for NK effector cells
Figure 5.Reproducibility of ADCC assay using cryopreserved NK cells from different donors. E:T ratio 6:1 (A) or E:T ratio 18:1 (B).
E:T ratio
Donor 1
Donor 2
Donor 3
Target cell killing
6:1
47.9%
43.1%
34.1%
18:1
57.8%
75.0%
55.0%
EC50 (anti-EGFR antibody)
6:1
1.1 ng/mL
3.9 ng/mL
5.3 ng/mL
18:1
1.2 ng/mL
5.6 ng/mL
4.2 ng/mL
Effects of Cryopreservation on Immune Cell Function
Cryopreserved PBMCs and immune cells offer a convenient, ready-to-use alternative to freshly isolated immune cells. We examined the performance of cryopreserved NK cells in ADCC assays. Though the ADCC assay required a longer incubation time (24 hours), robust cell killing and reproducible anti-EGFR antibody dose-response results were observed. Cryopreserved NK cells and PBMCs offer a practical alternative to freshly isolated immune cells for highly reproducible data in antibody-dependent cell-mediated cytotoxicity assays.
Frozen PBMC and Immune Cell Products
Sigma-Aldrich® is pleased to offer a wide range of PBMCs and immune cell subtypes from BioIVT.
Normal Human PBMCs
Product Description
Product Size
Product No.
Untyped
HLA typed
Frozen Human Normal PBMC
10 million cells per vial
HUMANPBMC-0001993
HUMANHLPB-0002560
20 million cells per vial
HUMANPBMC-0002144
HUMANHLPB-0002572
50 million cells per vial
HUMANPBMC-0002145
HUMANHLPB-0002056
100 million cells per vial
HUMANPBMC-0002644
HUMANHLPB-0002561
Immune Cell Subtypes
Product Description
Product Size
Product No.
Untyped
HLA typed
Frozen Human Normal Bone Marrow Mononuclear Cells (BMMCs)
10 million cells per vial
HUMANBMMC-0002115
HUMANHLBM-0002228
20 million cells per vial
HUMANBMMC-0002116
HUMANHLBM-0002229
50 million cells per vial
HUMANBMMC-0002117
HUMANHLBM-0002086
Frozen Human Normal CD14+ Monocytes
10 million cells per vial
HUMANCD14-0002081
HUMANHL14-0002136
20 million cells per vial
HUMANCD14-0002080
HUMANHL14-0002135
40 million cells per vial
HUMANCD14-0002079
HUMANHL14-0002059
Frozen Human Normal CD19+ B Cells
10 million cells per vial
HUMANCD19-0002069
HUMANHL19-0002057
Frozen Human Normal CD19+/IgD+ Naïve B Cells
1 million cells per vial
HUMANHLIG-0002064
Frozen Human Normal CD3+ Pan T Cells
10 million cells per vial
HUMANCD3-0002104
HUMANHLT3-0002227
20 million cells per vial
HUMANCD3-0002105
HUMANHLT3-0002072
Frozen Human Normal CD4+ Helper T Cells
10 million cells per vial
HUMANCD4-0002099
HUMANHLT4-0002226
Frozen Human Normal CD4+/CD25+ Regulatory T Cells
2 million cells per vial
HUMANTREG-0002094
HUMANHLTR-0002225
Frozen Human Normal CD4+/CD45RA+/CD25- Naive T Cells
5 million cells per vial
HUMANNCD4-0002653
HUMANHL4RA-0043279
Frozen Human Normal CD4+/CD45RO+ Memory T Cells
5 million cells per vial
HUMANMCD4-0002655
HUMANHL4RO-0002224
Frozen Human Normal CD8+ Cytotoxic T Cells
10 million cells per vial
HUMANCD8-0002096
HUMANHLT8-0002627
Frozen Human Normal CD8+/CD45RA+ Naïve Cytotoxic T Cells
5 million cells per vial
HUMANNCD8-0002090
HUMANHL8RA-0002498
Frozen Human Normal CD56+ NK Cells, positively selected
5 million cells per vial
HUMANCD56-0002665
HUMANHL56-0002058
Frozen Human Normal CD56+ NK Cells, negatively selected
5 million cells per vial
HUMANCD56-0002075
HUMANHL56-0002824
Frozen Human Normal CD34+ Hematopoietic Stem Cells
0.2 million cells per vial
HUMANCD34-0002046
HUMANHL34-0002236
1 million cells per vial
HUMANCD34-0002042
HUMANHL34-0002232
5 million cells per vial
HUMANCD34-0002043
HUMANHL34-0002233
10 million cells per vial
HUMANCD34-0002044
HUMANHL34-0002234
20 million cells per vial
HUMANCD34-0002045
HUMANHL34-0002235