Merck
LC-MS/MS Analysis of Melamine Extracted from Milk on Ascentis® Express HILIC
LC-MS/MS Analysis of Melamine Extracted from Milk on Ascentis® Express HILIC application for HPLC
材料
分析柱
产品编号
说明
价格
Ascentis® Express HILIC, 2.7 μm HPLC色谱柱
2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
標準品
产品编号
说明
价格
三聚氰胺
analytical standard
流動相成分
产品编号
说明
价格
甲酸铵
eluent additive for LC-MS, LiChropur, ≥99.0%
CONDITIONS
column
Ascentis Express HILIC, 10 cm x 2.1 mm I.D., 2.7 μm particles (53939-U)
mobile phase
[A] 10 mM ammonium formate in 10:90 (v/v) water:acetonitrile; [B] 10 mM ammonium formate in 30:70 (v/v) water:acetonitrile
gradient
held at 0% B for 5 min; 0 to 100% B in 0.1 min; held at 100% B for 4.9 min; 100 to 0% B in 0.1 min; held at 0% B for 4.9 min
flow rate
0.2 mL/min
column temp.
30 °C
detector
ABI 3200 Q-Trap
injection
2 μL
sample
SPE extract from spiked milk