Merck
HPLC Analysis of Ferredoxin NADP Reductase on TSKgel® Phenyl-5PW
HPLC Analysis of Ferredoxin NADP Reductase on TSKgel® Phenyl-5PW application for HPLC
材料
分析柱
产品编号
说明
价格
TSKgel®疏水作用色谱法(HIC)色谱柱
L × I.D. 7.5 cm × 7.5 mm, 10 μm particle size
CONDITIONS
column
TSKgel Phenyl-5PW column, 7.5 cm × 7.5 mm I.D., 10 μm particles (807573)
mobile phase
60 min decreasing linear gradient from 1.5M to 0M ammonium sulfate in 0.1M sodium phosphate buffer, pH7
flow rate
0.5 mL/min
column temp.
25 °C
detector
UV, 280 nm
injection
3mg (crude) or 0.2mg (standard)
说明
法律資訊
TSKgel is a registered trademark of Tosoh Corporation