返回
Hirschmann
®
Ceramus
®
bottle-top dispenser, adjustable volume
Products
产品分类
dispensers (6)
品牌
Sigma-Aldrich (6)
制造商
容量 (ml)
筛选产品
Showing 1-6 of 6
比较
货号
说明
价格
Z341495
Hirschmann
®
Ceramus
®
bottle-top dispenser, adjustable volume,
dispensing volume 2.0-10.0 mL, precision: 0.1 CV%
展开
查看定价
Z341509
Hirschmann
®
Ceramus
®
bottle-top dispenser, adjustable volume,
dispensing volume 5.0-30.0 mL, precision: 0.1 CV%
展开
查看定价
Z341487
Hirschmann
®
Ceramus
®
bottle-top dispenser, adjustable volume,
dispensing volume 1.0-5.0 mL, precision: 0.1 CV%
展开
查看定价
Z341517
Hirschmann
®
Ceramus
®
bottle-top dispenser, adjustable volume,
dispensing volume 10.0-60.0 mL, precision: 0.1 CV%
展开
查看定价
Z341479
Hirschmann
®
Ceramus
®
bottle-top dispenser, adjustable volume,
dispensing volume 0.4-2.0 mL, precision: 0.1 CV%
展开
查看定价
Z341460
Hirschmann
®
Ceramus
®
bottle-top dispenser, adjustable volume,
dispensing volume 0.2-1.0 mL, precision: 0.2 CV%