- Vitamin D
Vitamin D
Cellular immunology (2016-10-05)
L Malaguarnera, A Marsullo, K Zorena, G Musumeci, M Di Rosa
PMID27697285
ABSTRACT
The family of lysosome-associated membrane proteins (LAMPs) encompassing LAMP1, LAMP2 and DC-LAMP (LAMP3) are the major constituents of the glycoconjugates coat present on the inside of the lysosomal membrane. LAMP3 is highly expressed only in certain cell types and during the differentiation stages. Its expression is linked the maturation of dendritic cells, inflammation, poor prognosis of certain tumors, and the locus where it is encoded was identified as a risk factor for Parkinson's disease (PD). Here, we investigated the capacity of Vitamin D