Merck
  • Overexpression of karyopherin-α2 in cholangiocarcinoma correlates with poor prognosis and gemcitabine sensitivity via nuclear translocation of DNA repair proteins.
Overexpression of karyopherin-α2 in cholangiocarcinoma correlates with poor prognosis and gemcitabine sensitivity via nuclear translocation of DNA repair proteins.
Oncotarget (2017-02-09)
Mariko Tsukagoshi, Kenichiro Araki, Takehiko Yokobori, Bolag Altan, Hideki Suzuki, Norio Kubo, Akira Watanabe, Norihiro Ishii, Yasuo Hosouchi, Masahiko Nishiyama, Ken Shirabe, Hiroyuki Kuwano
ABSTRACT
Cholangiocarcinoma is a highly malignant tumor, and the development of new therapeutic strategies is critical. Karyopherin-α2 (KPNA2) functions as an adaptor that mediates nucleocytoplasmic transport. Specifically, KPNA2 transports one of the important DNA repair machineries, the MRE11-RAD50-NBS1 (MRN) complex, to the nucleus. In this study, we clarified the significance of KPNA2 in cholangiocarcinoma. KPNA2 expression evaluated by immunohistochemical analysis was common in malignant tissue but rare in adjacent noncancerous tissues. KPNA2 overexpression was significantly correlated with poor prognosis and was an independent prognostic factor after surgery. In patients with cholangiocarcinoma who received gemcitabine after surgery, KPNA2 overexpression tended to be a prognostic indicator of poor overall survival. In KPNA2-depleted cholangiocarcinoma cells, proliferation was significantly decreased and gemcitabine sensitivity was enhanced in vitro and in vivo. Expression of KPNA2 and the MRN complex displayed colocalization in the nucleus. In addition, nuclear localization of the MRN complex was regulated by KPNA2 in vitro. These results suggest that KPNA2 expression may be a useful prognostic and predictive marker of gemcitabine sensitivity and survival. The regulation of KPNA2 expression may be a new therapeutic strategy for cholangiocarcinoma.