Merck
  • Oriented coupling of major histocompatibility complex (MHC) to sensor surfaces using light assisted immobilisation technology.
Oriented coupling of major histocompatibility complex (MHC) to sensor surfaces using light assisted immobilisation technology.
Biosensors & bioelectronics (2005-09-06)
Torben Snabe, Gustav Andreas Røder, Maria Teresa Neves-Petersen, Søren Buus, Steffen Bjørn Petersen
ABSTRACT
Controlled and oriented immobilisation of proteins for biosensor purposes is of extreme interest since this provides more efficient sensors with a larger density of active binding sites per area compared to sensors produced by conventional immobilisation. In this paper oriented coupling of a major histocompatibility complex (MHC class I) to a sensor surface is presented. The coupling was performed using light assisted immobilisation--a novel immobilisation technology which allows specific opening of particular disulphide bridges in proteins which then is used for covalent bonding to thiol-derivatised surfaces via a new disulphide bond. Light assisted immobilisation specifically targets the disulphide bridge in the MHC-I molecule alpha(3)-domain which ensures oriented linking of the complex with the peptide binding site exposed away from the sensor surface. Structural analysis reveals that a similar procedure can be used for covalent immobilisation of MHC class II complexes. The results open for the development of efficient T cell sensors, sensors for recognition of peptides of pathogenic origin, as well as other applications that may benefit from oriented immobilisation of MHC proteins.