Merck
  • Molecular mechanisms of hyperthermia-induced apoptosis enhanced by withaferin A.
Molecular mechanisms of hyperthermia-induced apoptosis enhanced by withaferin A.
European journal of pharmacology (2013-12-11)
Zheng-Guo Cui, Jin-Lan Piao, Mati U R Rehman, Ryohei Ogawa, Peng Li, Qing-Li Zhao, Takashi Kondo, Hidekuni Inadera
ABSTRACT
Hyperthermia is a good therapeutic tool for non-invasive cancer therapy; however, its cytotoxic effects are not sufficient. In the present study, withaferin A (WA), a steroidal lactone derived from the plant Withania somnifera Dunal, has been investigated for its possible enhancing effects on hyperthermia-induced apoptosis. In HeLa cells, treatment with 0.5 or 1.0μM WA at 44°C for 30min induced significant apoptosis accompanied by decreased intracellular GSH/GSSG ratio and caspase-3 activation, while heat or WA alone did not induce such changes. The upregulation in apoptosis was significantly inhibited by glutathione monoethyl ester, a cell permeable glutathione precursor. Mitochondrial transmembrane potentials were dramatically decreased by the combined treatment, with increases in pro-apoptotic Bcl-2-family proteins tBid and Noxa, and downregulation of antiapoptotic Bcl-2 and Mcl-1. Combined treatment with hyperthermia and WA induced significant increases in JNK phosphorylation (p-JNK), and decreases in the phosphorylation of ERK (p-ERK) compared with either treatment alone. These results suggest that WA enhances hyperthermia-induced apoptosis via a mitochondria-caspase-dependent pathway; its underlying mechanism involves elevated intracellular oxidative stress, mitochondria dysfunction, and JNK activation.