Merck
  • The Tissue Fibrinolytic System Contributes to the Induction of Macrophage Function and CCL3 during Bone Repair in Mice.
The Tissue Fibrinolytic System Contributes to the Induction of Macrophage Function and CCL3 during Bone Repair in Mice.
PloS one (2015-04-22)
Naoyuki Kawao, Yukinori Tamura, Yoshitaka Horiuchi, Katsumi Okumoto, Masato Yano, Kiyotaka Okada, Osamu Matsuo, Hiroshi Kaji
ABSTRACT
Macrophages play crucial roles in repair process of various tissues. However, the details in the role of macrophages during bone repair still remains unknown. Herein, we examined the contribution of the tissue fibrinolytic system to the macrophage functions in bone repair after femoral bone defect by using male mice deficient in plasminogen (Plg-/-), urokinase-type plasminogen activator (uPA-/-) or tissue-type plasminogen activator (tPA-/-) genes and their wild-type littermates. Bone repair of the femur was delayed in uPA-/- mice until day 6, compared with wild-type (uPA+/+) mice. Number of Osterix-positive cells and vessel formation were decreased in uPA-/- mice at the bone injury site on day 4, compared with those in uPA+/+ mice. Number of macrophages and their phagocytosis at the bone injury site were reduced in uPA-/- and Plg-/-, but not in tPA-/- mice on day 4. Although uPA or plasminogen deficiency did not affect the levels of cytokines, including TNF-α, IL-1β, IL-6, IL-4 and IFN-γ mRNA in the damaged femur, the elevation in CCL3 mRNA levels was suppressed in uPA-/- and Plg-/-, but not in tPA-/- mice. Neutralization of CCL3 antagonized macrophage recruitment to the site of bone injury and delayed bone repair in uPA+/+, but not in uPA-/- mice. Our results provide novel evidence that the tissue fibrinolytic system contributes to the induction of macrophage recruitment and CCL3 at the bone injury site, thereby, leading to the enhancement of the repair process.